missing translation for 'onlineSavingsMsg'
Learn More

1-Butylhydantoin 98.0+%, TCI America™

Catalog No. b144825g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B144825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B144825G Supplier TCI America Supplier No. B144825G

Chemical Identifiers

CAS 33599-32-5
Molecular Formula C7H12N2O2
Molecular Weight (g/mol) 156.185
MDL Number MFCD00142674
InChI Key NOHNSECEKFOVGT-UHFFFAOYSA-N
Synonym 1-Butyl-2,4-dioxoimidazolidine
PubChem CID 118506
IUPAC Name 1-butylimidazolidine-2,4-dione
SMILES CCCCN1CC(=O)NC1=O

Specifications

Melting Point 94°C
Color White
Quantity 25 g
Formula Weight 156.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Butylhydantoin