missing translation for 'onlineSavingsMsg'
Learn More

1-Butylamine, 99%

Catalog No. p-7043940
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL03575AE 100 mL
AAL03575AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL03575AE Supplier Thermo Scientific Chemicals Supplier No. L03575AE
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 109-73-9 | C4H11N | 73.139 g/mol

1-Butylamine is used as an intermediate for pharmaceuticals, synthetic tanning agents, dyestuffs and emulsifiers. It also plays an important role in the preparation of N,N'-dibutylthiourea, a rubber vulcanization accelerator. It is involved in the preparation of n-butylbenzenesulfonamide, which is a plasticizer of nylon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Butylamine is used as an intermediate for pharmaceuticals, synthetic tanning agents, dyestuffs and emulsifiers. It also plays an important role in the preparation of N,N′-dibutylthiourea, a rubber vulcanization accelerator. It is involved in the preparation of n-butylbenzenesulfonamide, which is a plasticizer of nylon.

Solubility
Miscible with ethanol and water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, aluminum, copper, copper alloys and acids.

Chemical Identifiers

CAS 109-73-9
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.139
MDL Number MFCD00011690
InChI Key HQABUPZFAYXKJW-UHFFFAOYSA-N
Synonym butylamine, n-butylamine, 1-butanamine, 1-aminobutane, 1-butylamine, monobutylamine, n-butylamin, mono-n-butylamine, norvalamine, 1-aminobutan
PubChem CID 8007
ChEBI CHEBI:43799
IUPAC Name butan-1-amine
SMILES CCCCN

Specifications

Melting Point -50°C
Density 0.738
Boiling Point 76°C to 78°C
Flash Point −14°C (6°F)
Odor Ammonia-like
Refractive Index 1.401
Linear Formula CH3CH2CH2CH2NH2
Quantity 100 mL
UN Number UN1125
Beilstein 605269
Sensitivity Air Sensitive
Merck Index 14,1543
Solubility Information Miscible with ethanol and water.
Formula Weight 73.14
Percent Purity 99%
Chemical Name or Material 1-Butylamine
Show More Show Less
Hazard Category H225-H302-H311+H331-H314-H335
Hazard Statement GHS H Statement
H225-H314-H302-H312-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: N-BUTYLAMINE
EINECSNumber 203-699-2
RTECSNumber EO2975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only