missing translation for 'onlineSavingsMsg'
Learn More

(1-Butyl)triethylammonium bromide, 97%

Catalog No. AAB2412822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2412822 100 g
AAB2412814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2412822 Supplier Thermo Scientific Chemicals Supplier No. B2412822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13028-69-8 | C10H24BrN | 238.213 g/mol

(1-Butyl)triethylammonium bromide is an important organic intermediate. It can be used in agrochemical, pharmaceutical & dye stuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Butyl)triethylammonium bromide is an important organic intermediate. It can be used in agrochemical, pharmaceutical & dye stuff field.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, water/ moisture. Store under inert gas.

Chemical Identifiers

CAS 13028-69-8
Molecular Formula C10H24BrN
Molecular Weight (g/mol) 238.213
MDL Number MFCD00055541
InChI Key ZNFYPPCUDSCDPR-UHFFFAOYSA-M
Synonym 1-butyl triethylammonium bromide, butyltriethylammonium bromide, n,n,n-triethylbutan-1-aminium bromide, butyltriethylazanium bromide, acmc-20apri, triethylbutyl ammonium bromide, butyl-triethyl-ammonium bromide, triethyl-n-butylammonium bromide, 1-butanaminium,n,n,n-triethyl-, bromide 1:1
PubChem CID 11746670
IUPAC Name butyl(triethyl)azanium;bromide
SMILES CCCC[N+](CC)(CC)CC.[Br-]

Specifications

Melting Point 212°C to 215°C
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 238.21
Percent Purity 97%
Chemical Name or Material (1-Butyl)triethylammonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only