missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butyl-4-ethynylbenzene 97.0+%, TCI America™
$82.02 - $328.08
Chemical Identifiers
| CAS | 79887-09-5 |
|---|---|
| Molecular Formula | C12H14 |
| Molecular Weight (g/mol) | 158.24 |
| MDL Number | MFCD00173885 |
| InChI Key | ZVWWYEHVIRMJIE-UHFFFAOYSA-N |
| Synonym | 4-butylphenylacetylene, 1-butyl-4-eth-1-ynylbenzene, 1-butyl-4-eth1-ynylbenzene, 1-butyl-4-ethynyl-benzene, 4-n-butylphenylacetylene, p-butylphenylacetylene, benzene, 1-butyl-4-ethynyl, pubchem10978, acmc-209pic, p-butylphenyl acetylene |
| PubChem CID | 2775132 |
| IUPAC Name | 1-butyl-4-ethynylbenzene |
| SMILES | CCCCC1=CC=C(C=C1)C#C |
Chemical Identifiers
| 79887-09-5 | |
| 158.24 | |
| ZVWWYEHVIRMJIE-UHFFFAOYSA-N | |
| 2775132 | |
| CCCCC1=CC=C(C=C1)C#C |
| C12H14 | |
| MFCD00173885 | |
| 4-butylphenylacetylene, 1-butyl-4-eth-1-ynylbenzene, 1-butyl-4-eth1-ynylbenzene, 1-butyl-4-ethynyl-benzene, 4-n-butylphenylacetylene, p-butylphenylacetylene, benzene, 1-butyl-4-ethynyl, pubchem10978, acmc-209pic, p-butylphenyl acetylene | |
| 1-butyl-4-ethynylbenzene |
Specifications
| 79887-09-5 | |
| C12H14 | |
| 5 g | |
| ZVWWYEHVIRMJIE-UHFFFAOYSA-N | |
| 1-butyl-4-ethynylbenzene | |
| 2775132 | |
| ≥97.0% (GC) | |
| 1-Butyl-4-ethynylbenzene |
| Yellow | |
| MFCD00173885 | |
| 4-butylphenylacetylene, 1-butyl-4-eth-1-ynylbenzene, 1-butyl-4-eth1-ynylbenzene, 1-butyl-4-ethynyl-benzene, 4-n-butylphenylacetylene, p-butylphenylacetylene, benzene, 1-butyl-4-ethynyl, pubchem10978, acmc-209pic, p-butylphenyl acetylene | |
| CCCCC1=CC=C(C=C1)C#C | |
| 158.24 | |
| 158.24 | |
| Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator