missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-3-methylpyridinium Chloride 98.0+%, TCI America™

Catalog No. p-7122941
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34251G 1 g
B34255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34251G Supplier TCI America Supplier No. B34251G
Only null left

Chemical Identifiers

CAS 125652-55-3
Molecular Formula C10H16ClN
Molecular Weight (g/mol) 185.70
MDL Number MFCD08458942
InChI Key PHCASOSWUQOQAG-UHFFFAOYSA-M
PubChem CID 19876500
IUPAC Name 1-butyl-3-methylpyridin-1-ium chloride
SMILES [Cl-].CCCC[N+]1=CC=CC(C)=C1

Specifications

Quantity 1 g
Formula Weight 185.70
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Butyl-3-methylpyridinium Chloride