missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butyl-3-methylimidazolium Tetrachloroferrate 98.0+%, TCI America™
$348.57 - $1040.14
Chemical Identifiers
| CAS | 359845-21-9 |
|---|---|
| Molecular Formula | C8H15Cl4FeN2 |
| Molecular Weight (g/mol) | 336.867 |
| InChI Key | FYHLFBVHUIJIII-UHFFFAOYSA-J |
| PubChem CID | 11674455 |
| IUPAC Name | 1-butyl-3-methylimidazol-3-ium;tetrachloroiron(1-) |
| SMILES | CCCCN1C=C[N+](=C1)C.Cl[Fe-](Cl)(Cl)Cl |
Chemical Identifiers
| 359845-21-9 | |
| 336.867 | |
| 11674455 | |
| CCCCN1C=C[N+](=C1)C.Cl[Fe-](Cl)(Cl)Cl |
| C8H15Cl4FeN2 | |
| FYHLFBVHUIJIII-UHFFFAOYSA-J | |
| 1-butyl-3-methylimidazol-3-ium;tetrachloroiron(1-) |
Specifications
| 359845-21-9 | |
| C8H15Cl4FeN2 | |
| FYHLFBVHUIJIII-UHFFFAOYSA-J | |
| 1-butyl-3-methylimidazol-3-ium;tetrachloroiron(1-) | |
| 11674455 | |
| ≥98.0% (T) | |
| 1-Butyl-3-methylimidazolium Tetrachloroferrate |
| Green-Yellow | |
| 5 g | |
| CCCCN1C=C[N+](=C1)C.Cl[Fe-](Cl)(Cl)Cl | |
| 336.867 | |
| 336.87 | |
| Liquid |
Safety and Handling
TSCA : No