missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-3-methylimidazolium Methanesulfonate 98.0+%, TCI America™

Catalog No. B32315G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32315G 5 g
B323125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32315G Supplier TCI America Supplier No. B32315G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 342789-81-5
Molecular Formula C9H18N2O3S
Molecular Weight (g/mol) 234.314
MDL Number MFCD06798173
InChI Key PUHVBRXUKOGSBC-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium methanesulfonate, 1-butyl-3-methylimidazolium mesylate, acmc-20aowi, c4mim mesylate, basionics™ st 78, dsstox_cid_27888, dsstox_rid_82641, dsstox_gsid_47912, 1-butyl-3-methylimidazolium methylsulfonate, 1-n-butyl-3-methylimidazolium methanesulfonate
PubChem CID 11492381
IUPAC Name 1-butyl-3-methylimidazol-3-ium;methanesulfonate
SMILES CCCCN1C=C[N+](=C1)C.CS(=O)(=O)[O-]

Specifications

Melting Point 76°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 234.31
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Butyl-3-methylimidazolium Methanesulfonate