missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-3-methylimidazolium hexafluorophosphate, 98+%

CAS: 174501-64-5 | C8H15F6N2P | 284.19 g/mol

$309.20 - $309.20

Chemical Identifiers

CAS 174501-64-5
Molecular Formula C8H15F6N2P
Molecular Weight (g/mol) 284.19
MDL Number MFCD03093295
InChI Key IXQYBUDWDLYNMA-UHFFFAOYSA-N
Synonym 1-butyl-3-methylimidazolium hexafluorophosphate, 1-butyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, unii-zge3n4o8q9, 1-butyl-3-methylimidazoliumhexafluorophosphate, 1-n-butyl-3-methylimidazolium hexafluorophosphate, zge3n4o8q9, butylmethylimidazolium hexafluorophosphate, 1-methyl-3-butylimidazolium hexafluorophosphate, bmim pf6, c4mim hexafluorophosphate
PubChem CID 2734174
SMILES F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC354170250
SDS View Documents
Thermo Scientific Chemicals
354170250
25 g Glass bottle
Each for $309.20
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ionic liquid
Specifications

Chemical Identifiers

174501-64-5
284.19
IXQYBUDWDLYNMA-UHFFFAOYSA-N
2734174
C8H15F6N2P
MFCD03093295
1-butyl-3-methylimidazolium hexafluorophosphate, 1-butyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, unii-zge3n4o8q9, 1-butyl-3-methylimidazoliumhexafluorophosphate, 1-n-butyl-3-methylimidazolium hexafluorophosphate, zge3n4o8q9, butylmethylimidazolium hexafluorophosphate, 1-methyl-3-butylimidazolium hexafluorophosphate, bmim pf6, c4mim hexafluorophosphate
F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1

Specifications

174501-64-5
100.0
Colorless to Yellow
>340.0°C
98% min. (HPLC)
C8H15F6N2P
25 g
15,1584
IXQYBUDWDLYNMA-UHFFFAOYSA-N
1-butyl-3-methylimidazol-3-ium;hexafluorophosphate
2734174
284.18
Viscous Liquid
98.0
6.5°C
1.3727g/mL
Authentic
Glass bottle
MFCD03093295
1.3727
1-butyl-3-methylimidazolium hexafluorophosphate, 1-butyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, unii-zge3n4o8q9, 1-butyl-3-methylimidazoliumhexafluorophosphate, 1-n-butyl-3-methylimidazolium hexafluorophosphate, zge3n4o8q9, butylmethylimidazolium hexafluorophosphate, 1-methyl-3-butylimidazolium hexafluorophosphate, bmim pf6, c4mim hexafluorophosphate
F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1
284.19
272.1 mPa.s (25°C)
98+%
1-Butyl-3-methylimidazolium hexafluorophosphate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only