missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-3-methylimidazolium Chloride 98.0+%, TCI America™
GREENER_CHOICE

Catalog No. p-7119542
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Catalog No. Quantity
B21945G 5 g
B219425G 25 g
B2194100G 100 g
3 options

Catalog No. B21945G

Supplier: TCI America B21945G

Only null left

Chemical Identifiers

CAS 79917-90-1
Molecular Formula C8H15ClN2
Molecular Weight (g/mol) 174.67
MDL Number MFCD03095425
InChI Key FHDQNOXQSTVAIC-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium chloride, 3-butyl-1-methyl-1h-imidazol-3-ium chloride, 1-n-butyl-3-methylimidazolium chloride, bmimcl, bmim cl, c4mim chloride, dsstox_cid_11461, dsstox_rid_78878, dsstox_gsid_31461
PubChem CID 2734161
IUPAC Name 1-butyl-3-methyl-1H-imidazol-3-ium chloride
SMILES [Cl-].CCCCN1C=C[N+](C)=C1

Specifications

Quantity 5 g
UN Number 2811
Formula Weight 174.67
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Butyl-3-methylimidazolium Chloride