missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-2,3-dimethylimidazolium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. B24755G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24755G 5 g
B247525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B24755G Supplier TCI America Supplier No. B24755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 402846-78-0
Molecular Formula C9H17BF4N2
Molecular Weight (g/mol) 240.05
MDL Number MFCD03427618
InChI Key VCAIYEJBOWHUGP-UHFFFAOYSA-N
Synonym 1-butyl-2,3-dimethylimidazolium tetrafluoroborate, 1-n-butyl-2,3-dimethylimidazolium tetrafluoroborate, 1-butyl-2,3-dimethylimidazol-1-ium tetrafluoroborate, 3-butyl-1,2-dimethylimidazol-1-ium tetrafluoroborate, bdmim bf4, acmc-209jcq, dsstox_cid_27885, dsstox_rid_82638, dsstox_gsid_47909, ksc235m3b
PubChem CID 2734244
IUPAC Name 1-butyl-2,3-dimethyl-1H-imidazol-3-ium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1C

Specifications

Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 240.05
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Butyl-2,3-dimethylimidazolium Tetrafluoroborate