missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butyl-2,3-dimethylimidazolium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
$252.20 - $804.72
Chemical Identifiers
| CAS | 350493-08-2 |
|---|---|
| Molecular Formula | C11H17F6N3O4S2 |
| Molecular Weight (g/mol) | 433.384 |
| InChI Key | UCCKRVYTJPMHRO-UHFFFAOYSA-N |
| PubChem CID | 11212667 |
| IUPAC Name | bis(trifluoromethylsulfonyl)azanide;1-butyl-2,3-dimethylimidazol-3-ium |
| SMILES | CCCCN1C=C[N+](=C1C)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F |
Chemical Identifiers
| 350493-08-2 | |
| 433.384 | |
| 11212667 | |
| CCCCN1C=C[N+](=C1C)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F |
| C11H17F6N3O4S2 | |
| UCCKRVYTJPMHRO-UHFFFAOYSA-N | |
| bis(trifluoromethylsulfonyl)azanide;1-butyl-2,3-dimethylimidazol-3-ium |
Specifications
| 350493-08-2 | |
| C11H17F6N3O4S2 | |
| UCCKRVYTJPMHRO-UHFFFAOYSA-N | |
| bis(trifluoromethylsulfonyl)azanide;1-butyl-2,3-dimethylimidazol-3-ium | |
| 11212667 | |
| ≥98.0% (T) | |
| 1-Butyl-2,3-dimethylimidazolium Bis(trifluoromethanesulfonyl)imide |
| Yellow | |
| 5 g | |
| CCCCN1C=C[N+](=C1C)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F | |
| 433.384 | |
| 433.38 | |
| Liquid |
Safety and Handling
TSCA : No