missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-1-methylpyrrolidinium Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. p-7124121
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55685G 5 g
B556825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55685G Supplier TCI America Supplier No. B55685G

Chemical Identifiers

CAS 367522-96-1
Molecular Formula C10H20F3NO3S
Molecular Weight (g/mol) 291.33
MDL Number MFCD08458925
InChI Key WZJDNKTZWIOOJE-UHFFFAOYSA-M
Synonym 1-Butyl-1-methylpyrrolidinium Triflate
PubChem CID 12195864
IUPAC Name 1-butyl-1-methylpyrrolidin-1-ium trifluoromethanesulfonate
SMILES [O-]S(=O)(=O)C(F)(F)F.CCCC[N+]1(C)CCCC1

Specifications

Melting Point 4°C
Color Yellow
Quantity 5 g
Formula Weight 291.33
Percent Purity ≥98.0% (N)
Physical Form Liquid
Chemical Name or Material 1-Butyl-1-methylpyrrolidinium Trifluoromethanesulfonate