missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-1-methylpyrrolidinium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. B28515G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28515G 5 g
B285125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28515G Supplier TCI America Supplier No. B28515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 223437-11-4
Molecular Formula C11H20F6N2O4S2
Molecular Weight (g/mol) 422.40
MDL Number MFCD07784447
InChI Key HSLXOARVFIWOQF-UHFFFAOYSA-N
Synonym 1-butyl-1-methylpyrrolidinium bis trifluoromethylsulfonyl imide, unii-fd74dkk1wh, fd74dkk1wh, 1-butyl-1-methylpyrrolidinium bis trifluoromethanesulfonyl imide, 1-butyl-1-methylpyrrolidin-1-ium bis trifluoromethyl sulfonyl amide, 1-butyl-1-methylpyrrolidin-1-ium; bistriflylimide anion, dsstox_cid_27944, dsstox_rid_82696, dsstox_gsid_47968, pyrrolidinium, 1-butyl-1-methyl-, salt with 1,1,1-trifluoro-n-trifluoromethyl sulfonyl methanesulfonamide 1:1
PubChem CID 11048104
IUPAC Name 1-butyl-1-methylpyrrolidin-1-ium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
SMILES CCCC[N+]1(C)CCCC1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 422.40
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Butyl-1-methylpyrrolidinium Bis(trifluoromethanesulfonyl)imide