missing translation for 'onlineSavingsMsg'
Learn More

1-Butoxy-2-propanol 96.0+%, TCI America™

Catalog No. B0864500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0864500ML 500 mL
B086425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0864500ML Supplier TCI America Supplier No. B0864500ML
Only null left

Chemical Identifiers

CAS 5131-66-8
Molecular Formula C7H16O2
Molecular Weight (g/mol) 132.203
MDL Number MFCD00021904
InChI Key RWNUSVWFHDHRCJ-UHFFFAOYSA-N
Synonym Propylene Glycol 1-Monobutyl Ether
PubChem CID 21210
IUPAC Name 1-butoxypropan-2-ol
SMILES CCCCOCC(C)O

Specifications

Color Colorless
Boiling Point 170°C
Quantity 500 mL
Formula Weight 132.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Butoxy-2-propanol
EINECSNumber (2)-2424&(7)-0097
RTECSNumber UA7700000
TSCA Yes