missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butoxy-2-propanol 96.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 170°C |
| Quantity | 25 mL |
| Formula Weight | 132.20 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1-Butoxy-2-propanol |
Safety and Handling
| EINECSNumber | (2)-2424&(7)-0097 |
| RTECSNumber | UA7700000 |
| TSCA | Yes |