missing translation for 'onlineSavingsMsg'
Learn More

1-Buten-3-ol 97.0+%, TCI America™

Catalog No. B069525ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B069525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B069525ML Supplier TCI America Supplier No. B069525ML

missing translation for 'validMainframeMsg'

An unsaturated alcohol used as a chemical building block in synthesis.

Chemical Identifiers

CAS 598-32-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00004543
InChI Key MKUWVMRNQOOSAT-UHFFFAOYNA-N
Synonym 3-buten-2-ol, 1-buten-3-ol, methyl vinylcarbinol, 3-butene-2-ol, 3-hydroxy-1-butene, propenol, 1-methyl, methyl vinyl carbinol, 1-methyl-2-propenol, 2-methyl-2-propenol, 1-methylallyl alcohol
PubChem CID 11716
IUPAC Name but-3-en-2-ol
SMILES CC(O)C=C

Specifications

Color Yellow
Boiling Point 100°C
Quantity 25 mL
UN Number 1986
Formula Weight 72.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Buten-3-ol
EINECSNumber (2)-3171
RTECSNumber EM9275050
TSCA Yes