missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol, Ultrapure, Spectrophotometric Grade, 99.0+%

Catalog No. p-7049973
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
Catalog No. Quantity
AA32443K2 1 L
AA32443M1 4 x 1 L
AA32443K7 4 L
3 options

Catalog No. AA32443K2

Supplier: Thermo Scientific Chemicals 032443K2

Only null left

CAS: 71-36-3 | C4H10O | 74.123 g/mol

1-Butanol is primarily used as a solvent, as an intermediate in chemical synthesis, and as a fuel. It is also used as a paint thinner and a solvent in other coating applications where it is used as a relatively slow evaporating latent solvent in lacquers and ambient-cured enamels. It finds other uses such as a component of hydraulic and brake fluids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

pH 7
Melting Point -90°C
Color Colorless
Density 0.810 g/mL
Boiling Point 117°C to 118°C
Quantity 1 L
Flash Point 35°C (95°F)
Odor Alcohol-like
Physical Form Liquid
Refractive Index 1.399
Linear Formula CH3(CH2)3OH
UN Number UN1120
Beilstein 969148
Merck Index 14,1540
Solubility Information Soluble in water.
Formula Weight 74.12
Percent Purity ≥99.0%
Grade Ultrapure
Chemical Name or Material 1-Butanol
Show More Show Less
Hazard Category H226-H302-H315-H318-H335-H336
Hazard Statement GHS H Statement
H318-H226-H302-H335-H336-H315
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTANOLS
EINECSNumber 200-751-6
RTECSNumber EO1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only