missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol, for HPLC

Catalog No. p-3770952
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC393750010 1 L Glass Bottle
AC393750025 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC393750010 Supplier Thermo Scientific Chemicals Supplier No. 393750010
Only null left

CAS: 71-36-3 | C4H10O | 74.123 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point -89.0°C
Absorbance 0.046 max. (1cm cell vs HLPC−grade water) at 245nm, 0.301 max. (1cm cell vs HPLC−grade water) at 220nm, 0.699 max. (1 cm cell vs HPLC−grade water) at 210nm
Density 0.8100g/mL
Quantity 1 L
Flash Point 35°C
Packaging Glass Bottle
Refractive Index 1.3980 to 1.4000
Specific Gravity 0.81
Linear Formula CH3(CH2)3OH
Viscosity 2.95 mPa.s (20°C)
Boiling Point 117.6°C
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: miscible with ethanol, ether and many other, organic solvents
Formula Weight 74.12
Grade HPLC
Chemical Name or Material 1-Butanol, For HPLC
Show More Show Less
Hazard Category Danger

RUO – Research Use Only