missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5% (GC), Honeywell Riedel-de Haën™

Catalog No. p-7120026
Change view
Click to view available options
Quantity:
1 L
2.5 L
30 kg
4 x 2.5 L
6 x 1 L
Packaging:
Not available
Poly Bottle
Stainless Steel Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046183 1 L Poly Bottle
60046184 2.5 L Poly Bottle
60046187 6 x 1 L Poly Bottle
60046186 4 x 2.5 L Not available
60046185 30 kg Stainless Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60046183 Supplier Honeywell Chemicals Supplier No. 330651L
Only null left

Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99.5% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Color Colorless
Density 0.81g/mL at 25°C
Quantity 1 L
Explosion Limit 0.112
Flash Point 35°C (95°F)
Ignition Point 649°F
Packaging Poly Bottle
Refractive Index n20/D 1.399
Linear Formula CH3(CH2)3OH
UN Number UN1120
Beilstein 969148
Formula Weight 74.12g/mol
Percent Purity ≥99.5% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material 1-Butanol, Reag. Ph. Eur. reag. ISO
Show More Show Less
Hazard Category Danger
Hazard Statement P210-P280-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338 + P310-P403 + P235
Precautionary Statement H226-H302-H315-H318-H335-H336
DOTInformation Class 3 - PG 3 - Butanols
EINECSNumber 200-751-6
RTECSNumber EO1400000