missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol, CHROMASOLV™ Plus, for HPLC, ≥99.7%, Honeywell Riedel-de Haën™

Catalog No. 60046489
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60046489 6 x 1 L
6000087 1 L
6000088 2.5 L
6000090 4 x 2.5 L
60048234 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60046489 Supplier Honeywell Chemicals Supplier No. 348676X1L
Only null left

CHROMASOLV™ Plus, for HPLC,≥99.7%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point -90°C (literature)
Absorbance max. 0.10 at 240 nm, max. 0.10 at 235 nm, max. 0.50 at 225 nm, max. 0.70 at 220 nm, max. 1.0 at 210 nm, max. 0.01 at 400 nm, max. 0.01 280 nm, max. 0.04 at 260 nm
CAS Max % 100.0000%
Density 0.81 g/mL at 25 °C(lit.)
Quantity 6 x 1 L
Flash Point 35°C
Packaging Glass Bottle
Linear Formula C6H5CH3
MDL Number MFCD00008512
UN Number UN1120
Beilstein 969148
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
Boiling Point 119°C
Molecular Weight (g/mol) 74.123
Formula Weight 74.12
Percent Purity ≥99.7%
Grade HPLC
Chemical Name or Material 1-Butanol, CHROMASOLV™ Plus, for HPLC, ≥99.7%
Show More Show Less
ShelfLife 720 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Butanols
EINECSNumber 200-751-6
Recommended Storage Room Temp