missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol, CHROMASOLV™ Plus, for HPLC, ≥99.7%, Solstice

Catalog No. 6000087
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000087 1 L
6000088 2.5 L
60046489 6 x 1 L
6000090 4 x 2.5 L
60048234 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 6000087 Supplier Solstice Supplier No. 348671L

CHROMASOLV™ Plus, for HPLC,≥99.7%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point -90°C
Density 0.81g/mL at 25°C
Quantity 1 L
Refractive Index n20/D 1.399
Linear Formula CH3(CH2)3OH
MDL Number MFCD00002964
Beilstein 969148
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
Boiling Point 116°C to 118°C
Molecular Weight (g/mol) 74.123
Formula Weight 74.12g/mol
Percent Purity ≥99.7%
Grade CHROMASOLV™ Plus
Chemical Name or Material 1-Butanol, for HPLC
Show More Show Less
Hazard Statement P210-P280-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338 + P310-P403 + P235
Precautionary Statement H226-H302-H315-H318-H335-H336
EINECSNumber 200-751-6
RTECSNumber EO1400000