missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol (Certified ACS), Fisher Chemical™

Catalog No. p-215780
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A3994 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A3994 Supplier Thermo Fisher Scientific Supplier No. BPA3994
Only null left

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point -89°C
Color Colorless
Density 0.810g/cm³
Quantity 4 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.005% max.
Specific Gravity 0.806 to 0.810 (at 25°C)
Assay Percent Range ≥99.4 %
Titratable Acid 0.0008mEq/g max.
Vapor Pressure 6.7mbar at 20°C
Linear Formula CH3CH2CH2CH2OH
Viscosity 2.95 mPa/s at 20°C
UN Number 1120; Colorless
Water 0.1% max.
Boiling Point 117.6°C
Formula Weight 74.12
Percent Purity ≥99.4%
Grade Suitable for fluorometric analysis
Chemical Name or Material 1-Butanol
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Risk of serious damage to eyes. Irritating to eyes, respiratory system and skin. Vapors may cause drowsiness and dizziness. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:2
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid