missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. p-4824573
Change view
Click to view available options
Quantity:
250 mL
1 L
4 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41867AK 250 mL
AA41867K2 1 L
AA41867M1 4 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA41867AK Supplier Thermo Scientific Chemicals Supplier No. 041867AK
Only null left

CAS: 71-36-3 | C4H10O | 74.123 g/mol

1-Butanol is used as solvent for many organic reactions in pharmaceutical and chemical industry, as a solvent in coating industries and also used as a diluent/reactant in the manufacture of urea-formaldehyde and melamine-formaldehyde resins. It is used in the production of drugs, alkaloids, additive in floor cleaners, consumer products and stain removers. It is used as mobile phase in paper and thin layer chromatography. It is also used as an alternative for gasoline and diesel fuels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

pH 7
Melting Point -90°C to -89°C
Density 0.810
Boiling Point 117°C to 118°C
Quantity 250 mL
Flash Point 35°C (95°F)
Odor Alcohol-like
Packaging Resealable ChemSeal™ Bottle under Argon
Physical Form Liquid
Refractive Index 1.399
UN Number UN1120
Beilstein 969148
Merck Index 14,1540
Solubility Information Miscible with alcohols,ketones,aldehydes,ethers,aliphatic and aromatic hydrocarbons. Slightly miscible with water.
Formula Weight 74.12
Percent Purity 99.9%
Chemical Name or Material 1-Butanol, Anhydrous
Show More Show Less
Hazard Category H226-H302-H315-H318-H335-H336
Hazard Statement GHS H Statement
H318-H226-H302-H335-H336-H315;
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTANOLS
EINECSNumber 200-751-6
RTECSNumber EO1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only