missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol, 99+%, for spectroscopy

Catalog No. AC167691000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167691000 100 mL Glass Bottle
AC167695000 500 mL Glass Bottle
AC167690025 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC167691000 Supplier Thermo Scientific Chemicals Supplier No. 167691000

CAS: 71-36-3 | C4H10O | 74.123 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point -89.0°C
Absorbance 0.097 max. at 238nm, 1 max. at 205nm, 0.4 max. at 220nm, 0.1 max. at 240nm, 0.04 max. at 260nm, 0.02 max. at 280nm, 0.01 max. 330nm, 0.602 max. at 210nm, 0.046 max. at 245nm, 0.009 max. at 270nm, (1 cm cell vs HPLC-grade water), (1 cm cell vs HPLC-grade water) at 0nm
CAS Min % 99.0
CAS Max % 100.0
Density 0.8100g/mL
Quantity 100 mL
Flash Point 35°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3980 to 1.4000
Residue after Evaporation 0.0005% max.
Specific Gravity 0.81
Assay Percent Range 99+%
Linear Formula CH3(CH2)3OH
Viscosity 2.95 mPa.s (20°C)
UN Number 1120
Water 0.05% max. (Coulometric)
Beilstein 01, 367
Merck Index 15, 1542
Boiling Point 117.6°C
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: miscible with ethanol, ether and many other, organic solvents
Formula Weight 74.12
Percent Purity ≥99%
Grade Spectroscopy
Chemical Name or Material 1-Butanol
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause drowsiness or dizziness.
Causes serious eye damage.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-751-6
RTECSNumber EO1400000
TSCA TSCA

RUO – Research Use Only