Learn More
1-Butanol, 99.5%, ACS reagent, meets the requirements of Reag.Ph.Eur.
CAS: 71-36-3 | C4H10O | 74.123 g/mol
Supplier: Thermo Scientific Chemicals 423490025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1-Butanol | |
| 71-36-3 | |
| 100.0 | |
| 117.6°C | |
| 35°C | |
| Liquid | |
| 0.005% max. | |
| 0.81 | |
| C4H10O | |
| Vapor Pressure: 6.7mbar at 20°C | |
| 0.1% max. (K.F.) | |
| 15, 1542 | |
| Solubility in water: 80g/L (20°C). Other solubilities: miscible with ethanol, ether and many other, organic solvents | |
| CCCCO | |
| 74.123 | |
| CHEBI:28885 | |
| 99.5% |
| -89.0°C | |
| 99.4 | |
| 0.8100g/mL | |
| 2.5 L | |
| Glass Bottle | |
| 1.3980 to 1.4000 | |
| 99.5% | |
| 0.0008 meq/g max. | |
| CH3(CH2)3OH | |
| 2.95 mPa.s (20°C) | |
| 01, 367 | |
| 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane | |
| LRHPLDYGYMQRHN-UHFFFAOYSA-N | |
| butan-1-ol | |
| 263 | |
| 74.12 | |
| ACS Reagent |
Chemical Identifiers
| 71-36-3 | |
| 74.123 | |
| 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane | |
| CHEBI:28885 | |
| CCCCO |
| C4H10O | |
| LRHPLDYGYMQRHN-UHFFFAOYSA-N | |
| 263 | |
| butan-1-ol |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause drowsiness or dizziness.
Causes serious eye damage.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only