missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butanol, 99,5%, réactif ACS, répond aux exigences de la Reag.Ph.Eur.
Description
Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.
Spécifications
Spécifications
| Nom ou substance chimique | 1-Butanol |
| CAS % minimum | 99.4 |
| CAS % maximal | 100.0 |
| Densité | 0.8100g/mL |
| Quantité | 500 mL |
| Emballage | EcoSafPak™ Bouteille en verre |
| Forme physique | Liquide |
| Résidus après évaporation | 0,005% max. |
| Fourchette de pourcentages d’essai | 99.5% |
| Acide titrable | 0,0008 mEq/g max. |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement: Causes skin irritation. Harmful if swallowed. May cause drowsiness or dizziness. Causes serious eye damage. May cause respiratory irritation. Flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Usage réservé à la recherche