missing translation for 'onlineSavingsMsg'
Learn More

1-Butanesulfonic acid, sodium salt, 99+%, HPLC grade

Catalog No. p-3777748
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC343800050 5 g Glass bottle
AC343800250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC343800050 Supplier Thermo Scientific Chemicals Supplier No. 343800050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2386-54-1 | C4H9NaO3S | 160.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ion pair chromatography

Chemical Identifiers

CAS 2386-54-1
Molecular Formula C4H9NaO3S
Molecular Weight (g/mol) 160.16
MDL Number MFCD00007540
InChI Key XQCHMGAOAWZUPI-UHFFFAOYSA-M
Synonym sodium 1-butanesulfonate, sodium butane-1-sulfonate, 1-butanesulfonic acid sodium salt, sodium butylsulfonate, 1-butanesulfonic acid, sodium salt, sodium butane-1-sulphonate, sodium butanesulfonate, ipc-alks-4, 1-butanesulfonic acid, sodium salt 1:1, acmc-1cdot
PubChem CID 4096517
IUPAC Name sodium;butane-1-sulfonate
SMILES [Na+].CCCCS([O-])(=O)=O

Specifications

CAS Min % 99.0
Absorbance (5.5% aq. soln., pH 3) (1 cm cell), (5.5% aq. soln., pH 3) (1 cm cell) at 0nm, 0.04 max. at 250nm, 0.04 max. at 280nm, 0.1 max. at 220nm, 1 max. at 210nm
CAS Max % 100.0
Melting Point >300.0°C
Color White
Loss on Drying 1% max. (1 g, 105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3(CH2)3SO3Na
Quantity 5 g
Beilstein 04, 8
Solubility Information (10% in water) Clear to slightly hazy colorless
Formula Weight 160.17
Percent Purity 99+%
Grade HPLC
Physical Form Crystalline Powder
Chemical Name or Material 1-Butanesulfonic acid, sodium salt
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and
EINECSNumber 219-201-1

RUO – Research Use Only