missing translation for 'onlineSavingsMsg'
Learn More

(1-Bromovinyl)trimethylsilane 96.0+%, TCI America™

Catalog No. p-7119253
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17691G 1 g
B17695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17691G Supplier TCI America Supplier No. B17691G

Chemical Identifiers

CAS 13683-41-5
Molecular Formula C5H11BrSi
Molecular Weight (g/mol) 179.132
MDL Number MFCD00014927
InChI Key VVDJVCJVVHHCIB-UHFFFAOYSA-N
Synonym 1-bromovinyl trimethylsilane, 1-bromo-1-trimethylsilylethylene, 1-bromoethenyl trimethylsilane, 1-bromovinyl trimethyl silane, silane, 1-bromoethenyl trimethyl, acmc-1buto, 1-bromoethenyl trimethyl silane, alpha-trimethylsilylvinyl bromide, 1-bromoethenyl trimethyl-silane
PubChem CID 83660
IUPAC Name 1-bromoethenyl(trimethyl)silane
SMILES C[Si](C)(C)C(=C)Br

Specifications

Color Yellow
Boiling Point 128°C
Quantity 1 g
UN Number 1993
Formula Weight 179.13
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (1-Bromovinyl)trimethylsilane
TSCA No
Recommended Storage Refrigerator