missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1-Bromovinyl)trimethylsilane 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 128°C |
| Quantity | 5 g |
| UN Number | 1993 |
| Formula Weight | 179.13 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | (1-Bromovinyl)trimethylsilane |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |