missing translation for 'onlineSavingsMsg'
Learn More

1-Bromopinacolone, 97+%

Catalog No. AAA1065509
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1065509 10 g
AAA1065518 50 g
AAA1065530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1065509 Supplier Thermo Scientific Chemicals Supplier No. A1065509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5469-26-1 | C6H11BrO | 179.057 g/mol

1-Bromopinacolone is used as an iIntermediate for triazole compounds ( a five-membered ring structure compound contains three nitrogens and two carbons in the ring) which have biological activities such as antiviral, antibacterial, antifungal and antituberculous.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromopinacolone is used as an iIntermediate for triazole compounds ( a five-membered ring structure compound contains three nitrogens and two carbons in the ring) which have biological activities such as antiviral, antibacterial, antifungal and antituberculous.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases, heat and light.

Chemical Identifiers

CAS 5469-26-1
Molecular Formula C6H11BrO
Molecular Weight (g/mol) 179.057
MDL Number MFCD00000206
InChI Key SAIRZMWXVJEBMO-UHFFFAOYSA-N
Synonym 1-bromopinacolone, bromopinacolone, 1-bromo-3,3-dimethyl-2-butanone, 1-bromopinacolin, bromopinacolin, bromomethyl tert-butyl ketone, pivaloylmethyl bromide, tert-butyl bromomethyl ketone, 2-butanone, 1-bromo-3,3-dimethyl, omega-brompinakolin german
PubChem CID 21642
IUPAC Name 1-bromo-3,3-dimethylbutan-2-one
SMILES CC(C)(C)C(=O)CBr

Specifications

Melting Point -10°C
Density 1.333
Boiling Point 189°C to 192°C
Flash Point 75°C (167°F)
Odor Pungent
Refractive Index 1.4655
Quantity 10 g
UN Number UN2810
Beilstein 506485
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 179.06
Percent Purity ≥97%
Chemical Name or Material 1-Bromopinacolone
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H331-H318-H227-H315-H335
Toxic if inhaled.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 226-794-0
RTECSNumber EL7010000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only