missing translation for 'onlineSavingsMsg'
Learn More

1-Bromopinacolone, 93%

CAS: 5469-26-1 | C6H11BrO | 179.06 g/mol

$106.05 - $106.05

Chemical Identifiers

CAS 5469-26-1
Molecular Formula C6H11BrO
Molecular Weight (g/mol) 179.06
MDL Number MFCD00000206
InChI Key SAIRZMWXVJEBMO-UHFFFAOYSA-N
Synonym 1-bromopinacolone, bromopinacolone, 1-bromo-3,3-dimethyl-2-butanone, 1-bromopinacolin, bromopinacolin, bromomethyl tert-butyl ketone, pivaloylmethyl bromide, tert-butyl bromomethyl ketone, 2-butanone, 1-bromo-3,3-dimethyl, omega-brompinakolin german
PubChem CID 21642
IUPAC Name 1-bromo-3,3-dimethylbutan-2-one
SMILES CC(C)(C)C(=O)CBr
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC306410050
SDS View Documents
Thermo Scientific Chemicals
306410050
5 mL Glass bottle
Each for $106.05
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5469-26-1
179.06
SAIRZMWXVJEBMO-UHFFFAOYSA-N
21642
CC(C)(C)C(=O)CBr
C6H11BrO
MFCD00000206
1-bromopinacolone, bromopinacolone, 1-bromo-3,3-dimethyl-2-butanone, 1-bromopinacolin, bromopinacolin, bromomethyl tert-butyl ketone, pivaloylmethyl bromide, tert-butyl bromomethyl ketone, 2-butanone, 1-bromo-3,3-dimethyl, omega-brompinakolin german
1-bromo-3,3-dimethylbutan-2-one

Specifications

5469-26-1
100.0
Brown-Yellow to Yellow
189°C to 192°C
Authentic
Glass bottle
1.4656 to 1.4676
MFCD00000206
1.33
SAIRZMWXVJEBMO-UHFFFAOYSA-N
1-bromo-3,3-dimethylbutan-2-one
21642
93%
1-Bromopinacolone
91.0
-10°C
1.3300g/mL
75°C
91% min. (GC)
C6H11BrO
(CH3)3CCOCH2Br
5 mL
1-bromopinacolone, bromopinacolone, 1-bromo-3,3-dimethyl-2-butanone, 1-bromopinacolin, bromopinacolin, bromomethyl tert-butyl ketone, pivaloylmethyl bromide, tert-butyl bromomethyl ketone, 2-butanone, 1-bromo-3,3-dimethyl, omega-brompinakolin german
CC(C)(C)C(=O)CBr
179.06
179.06
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

EINECSNumber : 226-794-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only