missing translation for 'onlineSavingsMsg'
Learn More

1-Bromoperfluorooctane, 98+%

Catalog No. p-7026464
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2247006 5 g
AAB2247014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2247006 Supplier Thermo Scientific Chemicals Supplier No. B2247006
Only null left

CAS: 423-55-2 | C8BrF17 | 498.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromoperfluorooctane 1-Bromoheptadecafluorooctane (perfluorooctyl bromide) can be used as contrast agent in the vascular system for fluorine-19 magnetic resonance imaging or as synthetic oxygen carrier. It is used as an O2 carrier in some emulsions.

Solubility
Not miscible or difficult to mix in water.

Notes
Incompatible with oxidizing agents, acids, metals.

Chemical Identifiers

CAS 423-55-2
Molecular Formula C8BrF17
Molecular Weight (g/mol) 498.97
MDL Number MFCD00042082
InChI Key WTWWXOGTJWMJHI-UHFFFAOYSA-N
Synonym perflubron, 1-bromoperfluorooctane, perfluorooctyl bromide, 1-bromoheptadecafluorooctane, oxygent, perfluoro-n-octyl bromide, imagent gi, perfluorooctylbromide, liquivent, perfluoroctyl bromide
PubChem CID 9873
ChEBI CHEBI:38803
IUPAC Name 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctane
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br

Specifications

Melting Point 6°C
Density 1.93
Boiling Point 141°C to 143°C
Flash Point None
Refractive Index 1.305
Linear Formula CF3(CF2)7Br
Quantity 5 g
Beilstein 2016022
Merck Index 14,7156
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 498.98
Percent Purity ≥98%
Chemical Name or Material 1-Bromoperfluorooctane
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-028-4
RTECSNumber RG8700000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only