missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromooctadecane, 96%, Thermo Scientific™
Chemical Identifiers
| CAS | 112-89-0 |
|---|---|
| Molecular Formula | C18H37Br |
| Molecular Weight (g/mol) | 333.40 |
| MDL Number | MFCD00000231 |
| InChI Key | WSULSMOGMLRGKU-UHFFFAOYSA-N |
| Synonym | octadecyl bromide, stearyl bromide, octadecane, 1-bromo, n-octadecyl bromide, bromooctadecane, octadecylbromide, 1-bromoctadecane, n-octadecylbromide, 1-bromooctodecane, bromo-n-octadecane |
| PubChem CID | 8218 |
| IUPAC Name | 1-bromooctadecane |
| SMILES | CCCCCCCCCCCCCCCCCCBr |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC190021000
|
Thermo Scientific Chemicals
190021000 |
100g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC190025000
|
Thermo Scientific Chemicals
190025000 |
500g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC190020025
|
Thermo Scientific Chemicals
190020025 |
2.5kg | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 112-89-0 | |
| 333.40 | |
| WSULSMOGMLRGKU-UHFFFAOYSA-N | |
| 8218 | |
| CCCCCCCCCCCCCCCCCCBr |
| C18H37Br | |
| MFCD00000231 | |
| octadecyl bromide, stearyl bromide, octadecane, 1-bromo, n-octadecyl bromide, bromooctadecane, octadecylbromide, 1-bromoctadecane, n-octadecylbromide, 1-bromooctodecane, bromo-n-octadecane | |
| 1-bromooctadecane |
Specifications
| 112-89-0 | |
| 100.0 | |
| 214.0°C to 216.0°C (12.0mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4620 to 1.4640 | |
| MFCD00000231 | |
| 01,I,69 | |
| octadecyl bromide, stearyl bromide, octadecane, 1-bromo, n-octadecyl bromide, bromooctadecane, octadecylbromide, 1-bromoctadecane, n-octadecylbromide, 1-bromooctodecane, bromo-n-octadecane | |
| WSULSMOGMLRGKU-UHFFFAOYSA-N | |
| 1-bromooctadecane | |
| 8218 | |
| 96% |
| 95.0 | |
| 0.9700g/mL | |
| >110°C | |
| 95% min. (GC) | |
| C18H37Br | |
| CH3(CH2)17Br | |
| 100g | |
| 0.97 | |
| Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents | |
| CCCCCCCCCCCCCCCCCCBr | |
| 333.40 | |
| 333.38 | |
| 1-Bromooctadecane, 96% |
Safety and Handling
EINECSNumber : 204-013-4
TSCA : TSCA
RUO â Research Use Only