missing translation for 'onlineSavingsMsg'
Learn More

1-Bromonaphthalene 95.0+%, TCI America™

Catalog No. B061825G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B061825G 25 g
B0618100G 100 g
B0618500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B061825G Supplier TCI America Supplier No. B061825G

Chemical Identifiers

CAS 90-11-9
Molecular Formula C10H7Br
Molecular Weight (g/mol) 207.07
MDL Number MFCD00003868
InChI Key DLKQHBOKULLWDQ-UHFFFAOYSA-N
Synonym naphthalene, 1-bromo, 1-naphthyl bromide, alpha-bromonaphthalene, bromonaphthalene, alpha-naphthyl bromide, .alpha.-bromonaphthalene, 1-bromo-naphthalene, .alpha.-naphthyl bromide, naphthalene, bromo, bromonapthalene
PubChem CID 7001
IUPAC Name 1-bromonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2Br

Specifications

Melting Point -2°C
Color Yellow
Boiling Point 282°C
Quantity 25 g
Formula Weight 207.07
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromonaphthalene
RTECSNumber QJ1545000
TSCA Yes