missing translation for 'onlineSavingsMsg'
Learn More

1-Bromonaphthalene, 96%

Catalog No. p-75320
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107151000 100 mL Glass bottle
AC107155000 500 mL Glass bottle
AC107150025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC107151000 Supplier Thermo Scientific Chemicals Supplier No. 107151000
Only null left

CAS: 90-11-9 | C10H7Br | 207.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90-11-9 , 580-13-2
Molecular Weight (g/mol) 207.07
MDL Number MFCD00003868
InChI Key DLKQHBOKULLWDQ-UHFFFAOYSA-N
Synonym naphthalene, 1-bromo, 1-naphthyl bromide, alpha-bromonaphthalene, bromonaphthalene, alpha-naphthyl bromide, .alpha.-bromonaphthalene, 1-bromo-naphthalene, .alpha.-naphthyl bromide, naphthalene, bromo, bromonapthalene
PubChem CID 7001
IUPAC Name 1-bromonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2Br

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Brown-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Molecular Formula C10H7Br
Refractive Index 1.6560 to 1.6590 (20°C, 589nm)
Linear Formula C10H7Br
Quantity 100 mL
Beilstein 05, 547
Merck Index 15, 1436
Vapor Pressure 14.8hPa at 20°C
Formula Weight 207.07
Percent Purity ≥95%
Physical Form Liquid
Chemical Name or Material 1-Bromonaphthalene, 96%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-965-2
RTECSNumber QJ1545000
TSCA TSCA

RUO – Research Use Only