missing translation for 'onlineSavingsMsg'
Learn More

1-(Bromomethyl)isoquinoline hydrobromide, 97%, Thermo Scientific™

Catalog No. CC05108DE
Change view
Click to view available options
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
CC05108DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC05108DE Supplier Thermo Scientific Chemicals Supplier No. CC05108DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 337508-56-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C10H9Br2N
Quantity 5g
MDL Number MFCD03086096
Synonym 1-bromomethyl isoquinoline hydrobromide, 1-bromomethylisoquinoline hydrobromide, isoquinoline, 1-bromomethyl-, hydrobromide 1:1, 1-bromomethylisoquinoline hbr, 1-bromomethyl-isoquinolin hydrobromide, 1-bromomethyl-isoquinoline hydrobromide, isoquinoline, 1-bromomethyl-, hydrobromide
InChI Key RWSWRJTWPXSENX-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=CN=C2CBr.Br
IUPAC Name 1-(bromomethyl)isoquinoline;hydrobromide
Molecular Weight (g/mol) 302.997
PubChem CID 2776245
Formula Weight 303
Percent Purity 97%
Chemical Name or Material 1-(Bromomethyl)isoquinoline hydrobromide
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.