missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(Bromomethyl)-4-phenoxybenzene, ≥97%, Thermo Scientific™
Specifications
Specifications
| CAS | 36881-42-2 |
| CAS Min % | 96.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 97% |
| Molecular Formula | C13H11BrO |
| Quantity | 250mg |
| MDL Number | MFCD04038693 |
| Synonym | 1-bromomethyl-4-phenoxybenzene, 4-phenoxybenzyl bromide, 1-bromomethyl-4-phenoxy-benzene, benzene,1-bromomethyl-4-phenoxy, 4-phenyloxybenzylbromide, 4-bromomethyl-1-phenoxybenzene, benzene, 1-bromomethyl-4-phenoxy |
| InChI Key | CPIGBCFBFZSCQI-UHFFFAOYSA-N |
| SMILES | C1=CC=C(C=C1)OC2=CC=C(C=C2)CBr |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Harmful if swallowed. Causes serious eye damage. Harmful in contact with skin. Harmful if inhaled. Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |