missing translation for 'onlineSavingsMsg'
Learn More

1-Bromomethyl-2,2-difluorocyclopropane, 97%

Catalog No. AAH34305MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34305MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH34305MD Supplier Thermo Scientific Chemicals Supplier No. H34305MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 77613-65-1 | C4H5BrF2 | 170.985 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 77613-65-1
Molecular Formula C4H5BrF2
Molecular Weight (g/mol) 170.985
MDL Number MFCD06797749
InChI Key TUDJNSKRXIUOAJ-UHFFFAOYSA-N
Synonym 1-bromomethyl-2,2-difluorocyclopropane, 2-bromomethyl-1,1-difluorocyclopropane, 2,2-difluorocyclopropylmethyl bromide, 2,2-difluorocycloprop-1-yl methyl bromide, bromomethyl-2,2-difluorocyclopropane, 1-bromomethyl-2,2-difluoro-cyclopropane, 2-brornomethyl-1,1-difluorocyclopropane, cyclopropane,2-bromomethyl-1,1-difluoro, cyclopropane, 2-bromomethyl-1,1-difluoro, 2-bromomethyl-1,1-bis fluoranyl cyclopropane
PubChem CID 2761402
IUPAC Name 2-(bromomethyl)-1,1-difluorocyclopropane
SMILES C1C(C1(F)F)CBr

Specifications

Boiling Point 105°C to 107°C
Quantity 250 mg
UN Number UN1992
Formula Weight 170.98
Percent Purity 97%
Chemical Name or Material 1-Bromomethyl-2,2-difluorocyclopropane
Hazard Category H226-H302-H315-H319-H335
Hazard Statement GHS H Statement
H331-H226-H302-H312
Toxic if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only