missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-5-chloropentane, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 221210050

 View more versions of this product

Catalog No. AC221210050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

1-Bromo-5-chloropentane
97.5
1.4000g/mL
75°C
98%
C5H10BrCl
Cl(CH2)5Br
5g
1.4
Solubility in water: insoluble. Other solubilities: freely soluble in methanol,ethanol,chloroform,and ether
ClCCCCCBr
185.49
185.49
54512-75-3
100.0
210°C to 212°C
Authentic
Glass bottle
1.4826 to 1.4846
MFCD00001016
01,III,346
pentane, 1-bromo-5-chloro, 5-chloropentyl bromide, 5-bromopentyl chloride, 1-bromo-5-chloro-pentane, 1-brom-5-chlorpentan, 5-chlorobromopentane, bromo-5-chloropentane, pubchem15271, bromo-5-chloro-pentane, 5-bromo-1-chloropentane
PHHNNDKXQVKJEP-UHFFFAOYSA-N
1-bromo-5-chloropentane
96070
98%

Chemical Identifiers

54512-75-3
185.49
PHHNNDKXQVKJEP-UHFFFAOYSA-N
96070
ClCCCCCBr
C5H10BrCl
MFCD00001016
pentane, 1-bromo-5-chloro, 5-chloropentyl bromide, 5-bromopentyl chloride, 1-bromo-5-chloro-pentane, 1-brom-5-chlorpentan, 5-chlorobromopentane, bromo-5-chloropentane, pubchem15271, bromo-5-chloro-pentane, 5-bromo-1-chloropentane
1-bromo-5-chloropentane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 259-193-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only