missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-5-chloropentane, 98%

Catalog No. AC221211000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221211000 100 g Glass bottle
AC221210250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC221211000 Supplier Thermo Scientific Chemicals Supplier No. 221211000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 54512-75-3 | C5H10BrCl | 185.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54512-75-3
Molecular Formula C5H10BrCl
Molecular Weight (g/mol) 185.49
MDL Number MFCD00001016
InChI Key PHHNNDKXQVKJEP-UHFFFAOYSA-N
Synonym pentane, 1-bromo-5-chloro, 5-chloropentyl bromide, 5-bromopentyl chloride, 1-bromo-5-chloro-pentane, 1-brom-5-chlorpentan, 5-chlorobromopentane, bromo-5-chloropentane, pubchem15271, bromo-5-chloro-pentane, 5-bromo-1-chloropentane
PubChem CID 96070
IUPAC Name 1-bromo-5-chloropentane
SMILES ClCCCCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.4000g/mL
Boiling Point 210°C to 212°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4826 to 1.4846
Linear Formula Cl(CH2)5Br
Quantity 100 g
Beilstein 01,III,346
Specific Gravity 1.4
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in methanol,ethanol,chloroform,and ether
Formula Weight 185.49
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromo-5-chloropentane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 259-193-7

RUO – Research Use Only