missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-5-chloropentane, 98%

Catalog No. AAB2047909
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2047909 10 g
AAB2047918 50 g
AAB2047930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2047909 Supplier Thermo Scientific Chemicals Supplier No. B2047909
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 54512-75-3 | C5H10BrCl | 185.49 g/mol

1-Bromo-5-chloropentane, is used as an primary and secondary intermediate. It is also used as a intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-5-chloropentane, is used as an primary and secondary intermediate. It is also used as a intermediate in organic syntheses.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from strong oxidizing agents, strong acids, strong bases.

Chemical Identifiers

CAS 54512-75-3
Molecular Formula C5H10BrCl
Molecular Weight (g/mol) 185.49
MDL Number MFCD00001016
InChI Key PHHNNDKXQVKJEP-UHFFFAOYSA-N
Synonym pentane, 1-bromo-5-chloro, 5-chloropentyl bromide, 5-bromopentyl chloride, 1-bromo-5-chloro-pentane, 1-brom-5-chlorpentan, 5-chlorobromopentane, bromo-5-chloropentane, pubchem15271, bromo-5-chloro-pentane, 5-bromo-1-chloropentane
PubChem CID 96070
IUPAC Name 1-bromo-5-chloropentane
SMILES ClCCCCCBr

Specifications

Density 1.408
Boiling Point 210°C to 212°C
Flash Point 95°C (203°F)
Refractive Index 1.4836
Quantity 10 g
Beilstein 1732341
Solubility Information Insoluble in water.
Formula Weight 185.5
Percent Purity 98%
Chemical Name or Material 1-Bromo-5-chloropentane
Hazard Category H227-H315-H319-H335
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 259-193-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only