missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-phenylnaphthalene 98.0+%, TCI America™

Catalog No. p-7123957
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51051G 1 g
B51055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51051G Supplier TCI America Supplier No. B51051G
Only null left

Chemical Identifiers

CAS 59951-65-4
Molecular Formula C16H11Br
Molecular Weight (g/mol) 283.17
MDL Number MFCD17012346
InChI Key ZARGVWJSXZDKRE-UHFFFAOYSA-N
PubChem CID 12275025
IUPAC Name 1-bromo-4-phenylnaphthalene
SMILES BrC1=C2C=CC=CC2=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 77°C
Color White
Boiling Point 153°C
Quantity 1 g
Formula Weight 283.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-4-phenylnaphthalene