missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-4-methoxybutane 98.0+%, TCI America™
$91.33 - $274.02
Chemical Identifiers
| CAS | 4457-67-4 |
|---|---|
| Molecular Formula | C5H11BrO |
| Molecular Weight (g/mol) | 167.046 |
| MDL Number | MFCD00037102 |
| InChI Key | ALOQTNHQNMYBDE-UHFFFAOYSA-N |
| Synonym | 4-Bromobutyl Methyl Ether |
| PubChem CID | 521168 |
| IUPAC Name | 1-bromo-4-methoxybutane |
| SMILES | COCCCCBr |
Chemical Identifiers
| 4457-67-4 | |
| 167.046 | |
| ALOQTNHQNMYBDE-UHFFFAOYSA-N | |
| 521168 | |
| COCCCCBr |
| C5H11BrO | |
| MFCD00037102 | |
| 4-Bromobutyl Methyl Ether | |
| 1-bromo-4-methoxybutane |
Specifications
| 4457-67-4 | |
| 165°C | |
| MFCD00037102 | |
| 1993 | |
| ALOQTNHQNMYBDE-UHFFFAOYSA-N | |
| 1-bromo-4-methoxybutane | |
| 521168 | |
| ≥98.0% (GC) | |
| 1-Bromo-4-methoxybutane |
| Colorless | |
| C5H11BrO | |
| 5 g | |
| 4-Bromobutyl Methyl Ether | |
| COCCCCBr | |
| 167.046 | |
| 167.05 | |
| Liquid |
Safety and Handling
TSCA : No