missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-4-iodonaphthalene 98.0+%, TCI America™
$155.93 - $621.84
Chemical Identifiers
| CAS | 63279-58-3 |
|---|---|
| Molecular Formula | C10H6BrI |
| Molecular Weight (g/mol) | 332.966 |
| MDL Number | MFCD01166242 |
| InChI Key | HQHHKYXPFKHLBF-UHFFFAOYSA-N |
| Synonym | 1-iodo-4-bromonaphthalene, 1-iodo-4-bromoaphthalene, 1-bromo-4-iodonaphth, naphthalene, 1-bromo-4-iodo |
| PubChem CID | 5073667 |
| IUPAC Name | 1-bromo-4-iodonaphthalene |
| SMILES | C1=CC=C2C(=C1)C(=CC=C2I)Br |
Chemical Identifiers
| 63279-58-3 | |
| 332.966 | |
| HQHHKYXPFKHLBF-UHFFFAOYSA-N | |
| 5073667 | |
| C1=CC=C2C(=C1)C(=CC=C2I)Br |
| C10H6BrI | |
| MFCD01166242 | |
| 1-iodo-4-bromonaphthalene, 1-iodo-4-bromoaphthalene, 1-bromo-4-iodonaphth, naphthalene, 1-bromo-4-iodo | |
| 1-bromo-4-iodonaphthalene |
Specifications
| 63279-58-3 | |
| C10H6BrI | |
| 1 g | |
| HQHHKYXPFKHLBF-UHFFFAOYSA-N | |
| 1-bromo-4-iodonaphthalene | |
| 5073667 | |
| ≥98.0% (GC) | |
| 1-Bromo-4-iodonaphthalene |
| 89°C | |
| MFCD01166242 | |
| 1-iodo-4-bromonaphthalene, 1-iodo-4-bromoaphthalene, 1-bromo-4-iodonaphth, naphthalene, 1-bromo-4-iodo | |
| C1=CC=C2C(=C1)C(=CC=C2I)Br | |
| 332.966 | |
| 332.97 | |
| Crystalline Powder |
Safety and Handling
TSCA : No