missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-iodonaphthalene 98.0+%, TCI America™

Catalog No. B48641G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48641G 1 g
B48645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48641G Supplier TCI America Supplier No. B48641G

Chemical Identifiers

CAS 63279-58-3
Molecular Formula C10H6BrI
Molecular Weight (g/mol) 332.966
MDL Number MFCD01166242
InChI Key HQHHKYXPFKHLBF-UHFFFAOYSA-N
Synonym 1-iodo-4-bromonaphthalene, 1-iodo-4-bromoaphthalene, 1-bromo-4-iodonaphth, naphthalene, 1-bromo-4-iodo
PubChem CID 5073667
IUPAC Name 1-bromo-4-iodonaphthalene
SMILES C1=CC=C2C(=C1)C(=CC=C2I)Br

Specifications

Melting Point 89°C
Quantity 1 g
Formula Weight 332.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-4-iodonaphthalene