missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-iodonaphthalene, 98%

Catalog No. p-7056077
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6482503 1 g
AAH6482506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6482503 Supplier Thermo Scientific Chemicals Supplier No. H6482503

CAS: 63279-58-3 | C10H6BrI | 332.966 g/mol

It is used as an OLED material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an OLED material.

Solubility
Slightly soluble in water. (3.6 g/L) (25°C)

Notes
Light sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 63279-58-3
Molecular Formula C10H6BrI
Molecular Weight (g/mol) 332.966
MDL Number MFCD01166242
InChI Key HQHHKYXPFKHLBF-UHFFFAOYSA-N
Synonym 1-iodo-4-bromonaphthalene, 1-iodo-4-bromoaphthalene, 1-bromo-4-iodonaphth, naphthalene, 1-bromo-4-iodo
PubChem CID 5073667
IUPAC Name 1-bromo-4-iodonaphthalene
SMILES C1=CC=C2C(=C1)C(=CC=C2I)Br

Specifications

Melting Point 84°C to 89°C
Quantity 1 g
Beilstein 2044950
Sensitivity Light sensitive
Solubility Information Slightly soluble in water. (3.6g/L) (25°C)
Formula Weight 332.97
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-Bromo-4-iodonaphthalene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only