Learn More
1-Bromo-4-fluorobutane, 97%
CAS: 462-72-6 | C4H8BrF | 155.01 g/mol
Supplier: Thermo Scientific Chemicals H6431706
Description
1-Bromo-4-fluorobutane may be employed for the synthesis of fluoroalkyl[2,5-dimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-yl]alkylamines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Bromo-4-fluorobutane may be employed for the synthesis of fluoroalkyl[2,5-dimethyl-7-(2,4,6-trimethylphenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-yl]alkylamines.
Solubility
Sparingly soluble in water at 25°C 0.48 g/L.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| 1-Bromo-4-fluorobutane | |
| Colorless | |
| 134°C to 135°C | |
| C4H8BrF | |
| FCH2CH2CH2CH2Br | |
| 5 g | |
| 1731414 | |
| Sparingly soluble in water at 25°C 0.48g/L. | |
| C(CCBr)CF | |
| 155.01 | |
| 155.01 | |
| Liquid |
| 462-72-6 | |
| 1.444 g/mL | |
| 45°C (113°F) | |
| 1.439 | |
| MFCD00209551 | |
| UN1993 | |
| 4-fluorobutyl bromide, butane, 1-bromo-4-fluoro, butane,1-bromo-4-fluoro, fluorobutyl bromide, acmc-20aoww, 4-bromo-1-fluorobutane, 1-bromo-4-fluoro-butane, 4-01-00-00263 beilstein handbook reference, 1-bromo-4-fluorobutane, 1-bromo-4-fluoro-butan;4-fluoro-1-bromobutane | |
| WMCUHRDQSHQNRW-UHFFFAOYSA-N | |
| 1-bromo-4-fluorobutane | |
| 10022 | |
| 97% |
Chemical Identifiers
| 462-72-6 | |
| 155.01 | |
| WMCUHRDQSHQNRW-UHFFFAOYSA-N | |
| 1-bromo-4-fluorobutane |
| C4H8BrF | |
| MFCD00209551 | |
| 10022 | |
| C(CCBr)CF |
Safety and Handling
GHS H Statement
H226-H302-H312-H332-H315-H319-H335
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
RTECSNumber : EJ6229000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only