missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-4-chlorobutane 99.0+%, TCI America™
$70.84 - $445.51
Chemical Identifiers
| CAS | 6940-78-9 |
|---|---|
| Molecular Formula | C4H8BrCl |
| Molecular Weight (g/mol) | 171.462 |
| MDL Number | MFCD00001010 |
| InChI Key | NIDSRGCVYOEDFW-UHFFFAOYSA-N |
| Synonym | butane, 1-bromo-4-chloro, tetramethylene chlorobromide, 4-bromo-1-chlorobutane, 1-chloro-4-bromobutane, 4-bromobutyl chloride, 4-chlorobutyl bromide, 1-brom-4-chlorbutan, 4-chlorobromobutane, bromo 4 chlorobutane, bromo-4-chlorobutane |
| PubChem CID | 81364 |
| IUPAC Name | 1-bromo-4-chlorobutane |
| SMILES | C(CCBr)CCl |
Chemical Identifiers
| 6940-78-9 | |
| 171.462 | |
| NIDSRGCVYOEDFW-UHFFFAOYSA-N | |
| 81364 | |
| C(CCBr)CCl |
| C4H8BrCl | |
| MFCD00001010 | |
| butane, 1-bromo-4-chloro, tetramethylene chlorobromide, 4-bromo-1-chlorobutane, 1-chloro-4-bromobutane, 4-bromobutyl chloride, 4-chlorobutyl bromide, 1-brom-4-chlorbutan, 4-chlorobromobutane, bromo 4 chlorobutane, bromo-4-chlorobutane | |
| 1-bromo-4-chlorobutane |
Specifications
| 6940-78-9 | |
| 81°C | |
| MFCD00001010 | |
| 1993 | |
| NIDSRGCVYOEDFW-UHFFFAOYSA-N | |
| 1-bromo-4-chlorobutane | |
| 81364 | |
| ≥99.0% (GC) | |
| 1-Bromo-4-chlorobutane |
| Yellow | |
| C4H8BrCl | |
| 25 g | |
| butane, 1-bromo-4-chloro, tetramethylene chlorobromide, 4-bromo-1-chlorobutane, 1-chloro-4-bromobutane, 4-bromobutyl chloride, 4-chlorobutyl bromide, 1-brom-4-chlorbutan, 4-chlorobromobutane, bromo 4 chlorobutane, bromo-4-chlorobutane | |
| C(CCBr)CCl | |
| 171.462 | |
| 171.46 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-3318
TSCA : Yes