missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-chlorobutane 99.0+%, TCI America™

Catalog No. B134525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B134525G 25 g
B1345500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B134525G Supplier TCI America Supplier No. B134525G

Chemical Identifiers

CAS 6940-78-9
Molecular Formula C4H8BrCl
Molecular Weight (g/mol) 171.462
MDL Number MFCD00001010
InChI Key NIDSRGCVYOEDFW-UHFFFAOYSA-N
Synonym butane, 1-bromo-4-chloro, tetramethylene chlorobromide, 4-bromo-1-chlorobutane, 1-chloro-4-bromobutane, 4-bromobutyl chloride, 4-chlorobutyl bromide, 1-brom-4-chlorbutan, 4-chlorobromobutane, bromo 4 chlorobutane, bromo-4-chlorobutane
PubChem CID 81364
IUPAC Name 1-bromo-4-chlorobutane
SMILES C(CCBr)CCl

Specifications

Color Yellow
Boiling Point 81°C
Quantity 25 g
UN Number 1993
Formula Weight 171.46
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-chlorobutane
EINECSNumber (2)-3318
TSCA Yes