missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-chlorobutane, 99%

CAS: 6940-78-9 | C4H8BrCl | 171.462 g/mol

$88.62 - $233.01

Chemical Identifiers

CAS 6940-78-9
Molecular Formula C4H8BrCl
Molecular Weight (g/mol) 171.462
MDL Number MFCD00001010
InChI Key NIDSRGCVYOEDFW-UHFFFAOYSA-N
Synonym butane, 1-bromo-4-chloro, tetramethylene chlorobromide, 4-bromo-1-chlorobutane, 1-chloro-4-bromobutane, 4-bromobutyl chloride, 4-chlorobutyl bromide, 1-brom-4-chlorbutan, 4-chlorobromobutane, bromo 4 chlorobutane, bromo-4-chlorobutane
PubChem CID 81364
IUPAC Name 1-bromo-4-chlorobutane
SMILES C(CCBr)CCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1139914
View Documents
Thermo Scientific Chemicals
A1139914
25 g
Each for $88.62
Only null left
 
AAA1139922
View Documents
Thermo Scientific Chemicals
A1139922
100 g
Each for $233.01
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6940-78-9
171.462
NIDSRGCVYOEDFW-UHFFFAOYSA-N
81364
C(CCBr)CCl
C4H8BrCl
MFCD00001010
butane, 1-bromo-4-chloro, tetramethylene chlorobromide, 4-bromo-1-chlorobutane, 1-chloro-4-bromobutane, 4-bromobutyl chloride, 4-chlorobutyl bromide, 1-brom-4-chlorbutan, 4-chlorobromobutane, bromo 4 chlorobutane, bromo-4-chlorobutane
1-bromo-4-chlorobutane

Specifications

6940-78-9
175°C to 176°C
Aromatic
1.4875
25 g
1098275
NIDSRGCVYOEDFW-UHFFFAOYSA-N
1-bromo-4-chlorobutane
81364
99%
1.489
60°C (140°F)
C4H8BrCl
MFCD00001010
UN1993
butane, 1-bromo-4-chloro, tetramethylene chlorobromide, 4-bromo-1-chlorobutane, 1-chloro-4-bromobutane, 4-bromobutyl chloride, 4-chlorobutyl bromide, 1-brom-4-chlorbutan, 4-chlorobromobutane, bromo 4 chlorobutane, bromo-4-chlorobutane
C(CCBr)CCl
171.462
171.47
1-Bromo-4-chlorobutane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 230-089-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only